Product Details

SNP ID
rs112599042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57625442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGATGTCGAGATGCTAGGATCCG[C/G]CTCCTCCTGGCTCAGTGTAATGGTG
Phenotype
MIM: 610215 MIM: 601873
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF25 PubMed Links

Gene Details

Gene
ARHGEF25
Gene Name
Rho guanine nucleotide exchange factor 25
There are no transcripts associated with this gene.

Gene
B4GALNT1
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276468.1 1817 Intron NP_001263397.1
NM_001276469.1 1817 Intron NP_001263398.1
NM_001478.4 1817 Intron NP_001469.1
XM_005268773.4 1817 UTR 3 XP_005268830.1
XM_011538147.2 1817 UTR 3 XP_011536449.1
XM_011538148.2 1817 UTR 3 XP_011536450.1
XM_017019140.1 1817 UTR 3 XP_016874629.1
XM_017019141.1 1817 UTR 3 XP_016874630.1
XM_017019142.1 1817 UTR 3 XP_016874631.1
Gene
LOC101927583
Gene Name
uncharacterized LOC101927583
There are no transcripts associated with this gene.

Gene
SLC26A10
Gene Name
solute carrier family 26 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133489.2 1817 Silent Mutation CGC,CGG R502R NP_597996.2

View Full Product Details