Product Details
- SNP ID
-
rs117074911
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71692854 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTAAAAACCTTTGCTTTTTTCCT[A/G]ATTATTTCATTCTATGAACTGTTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM19
PubMed Links
Gene Details
- Gene
- TMEM19
- Gene Name
- transmembrane protein 19
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018279.3 |
|
Intron |
|
|
NP_060749.2 |
View Full Product Details