Product Details

SNP ID
rs114397456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:74540126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTGTAGAAGACTCAGAGGAGA[A/G]TCTTTCTTATGGCTCCCTCTGTTGA
Phenotype
MIM: 615579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN7L3B PubMed Links

Gene Details

Gene
ATXN7L3B
Gene Name
ataxin 7 like 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136262.1 2356 UTR 3 NP_001129734.1

View Full Product Details