Product Details

SNP ID
rs115759085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92702923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAGGCTTTTTCATAAACAAATA[A/C]CTGGAAATGTCTCTAAGGAGGGGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C12orf74 PubMed Links

Gene Details

Gene
C12orf74
Gene Name
chromosome 12 open reading frame 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037671.3 81 UTR 5 NP_001032760.1
NM_001178097.2 81 UTR 5 NP_001171568.1

View Full Product Details