Product Details

SNP ID
rs112412762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93390228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGAATTGTGTAACAAGAATTT[C/T]ATCTAGGCAATAGAAACCAGTTTAT
Phenotype
MIM: 609229
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT4 PubMed Links

Gene Details

Gene
NUDT4
Gene Name
nudix hydrolase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301022.1 Intron NP_001287951.1
NM_001301023.1 Intron NP_001287952.1
NM_001301024.1 Intron NP_001287953.1
NM_019094.5 Intron NP_061967.3
NM_199040.3 Intron NP_950241.1
XM_011537794.2 Intron XP_011536096.1
XM_011537795.2 Intron XP_011536097.1
XM_011537796.2 Intron XP_011536098.1

View Full Product Details