Product Details
- SNP ID
-
rs112162917
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:99648394 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCGAATGGTAACAATGGGCATC[A/G]TTCGCACCAGCCCCTGCCTCACACT
- Phenotype
-
MIM: 607815
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKS1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11109969] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKS1B
- Gene Name
- ankyrin repeat and sterile alpha motif domain containing 1B
- Gene
- FAM71C
- Gene Name
- family with sequence similarity 71 member C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153364.3 |
645 |
Missense Mutation |
ATT,GTT |
I74V |
NP_699195.1 |
View Full Product Details