Product Details

SNP ID
rs115125002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100277265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCATTTGTAGAGTGTGTGTTTT[A/G]GGTCAGGTTGTTTGATAGTGCTGTT
Phenotype
MIM: 616365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEPDC4 PubMed Links

Gene Details

Gene
DEPDC4
Gene Name
DEP domain containing 4
There are no transcripts associated with this gene.

Gene
SCYL2
Gene Name
SCY1 like pseudokinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317784.1 Intron NP_001304713.1
NM_017988.5 Intron NP_060458.3
XM_017019618.1 Intron XP_016875107.1
XM_017019619.1 Intron XP_016875108.1
XM_017019620.1 Intron XP_016875109.1

View Full Product Details