Product Details

SNP ID
rs117368247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104986407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGTTGTTCGTCGCCCACCCGG[C/G]ATTCCTCAGGAGTCCCAGTGTCCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C12orf45 PubMed Links

Gene Details

Gene
C12orf45
Gene Name
chromosome 12 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152318.2 88 Missense Mutation CAT,GAT H19D NP_689531.2

View Full Product Details