Product Details

SNP ID
rs117136616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:45220666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAGGCATTCTTCCAGTTTCTTG[G/T]CATGCAACTCCTAAAACTTTTGGAA
Phenotype
MIM: 608663
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANO6 PubMed Links

Gene Details

Gene
ANO6
Gene Name
anoctamin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025356.2 Intron NP_001020527.2
NM_001142678.1 Intron NP_001136150.1
NM_001142679.1 Intron NP_001136151.1
NM_001204803.1 Intron NP_001191732.1
XM_005268706.4 Intron XP_005268763.1
XM_005268707.3 Intron XP_005268764.1
Gene
PLEKHA8P1
Gene Name
pleckstrin homology domain containing A8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details