Product Details

SNP ID
rs117946594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196507588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGATATGTAAACTAATATGTG[C/T]TATATATATATTTATATATATATAT
Phenotype
MIM: 612688
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF168 PubMed Links

Gene Details

Gene
RNF168
Gene Name
ring finger protein 168
There are no transcripts associated with this gene.

Gene
SMCO1
Gene Name
single-pass membrane protein with coiled-coil domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077657.2 1101 UTR 3 NP_001071125.1
NM_001320473.1 1101 UTR 3 NP_001307402.1

View Full Product Details