Product Details

SNP ID
rs115541049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196642562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCCATGAGTTCTAGCAACTGC[C/T]CGTGGATTGATTCTCATGGGCTTGG
Phenotype
MIM: 615322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01063 PubMed Links

Gene Details

Gene
LINC01063
Gene Name
long intergenic non-protein coding RNA 1063
There are no transcripts associated with this gene.

Gene
NRROS
Gene Name
negative regulator of reactive oxygen species
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198565.2 Intron NP_940967.1

View Full Product Details