Product Details

SNP ID
rs111989729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110579622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAAGGGAGAAGAACTACACCT[C/T]TGGGCTGACTGCACAGGGGACAGTC
Phenotype
MIM: 609568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP20 PubMed Links

Gene Details

Gene
ARHGAP20
Gene Name
Rho GTPase activating protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258415.1 3430 Silent Mutation CAA,CAG Q1085Q NP_001245344.1
NM_001258416.1 3430 Silent Mutation CAA,CAG Q1082Q NP_001245345.1
NM_001258417.1 3430 Silent Mutation CAA,CAG Q1072Q NP_001245346.1
NM_001258418.1 3430 Silent Mutation CAA,CAG Q1072Q NP_001245347.1
NM_020809.3 3430 Silent Mutation CAA,CAG Q1108Q NP_065860.2
XM_005271628.3 3430 Silent Mutation CAA,CAG Q651Q XP_005271685.1
XM_006718893.3 3430 Silent Mutation CAA,CAG Q1108Q XP_006718956.1

View Full Product Details