Product Details

SNP ID
rs116006647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117330258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACATTTTGTAGTCATGTGTTTA[C/T]GTGTTTGGCTTTCCAGGTAGGCTGA
Phenotype
MIM: 614848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP164 PubMed Links

Gene Details

Gene
CEP164
Gene Name
centrosomal protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271933.1 Intron NP_001258862.1
NM_014956.4 Intron NP_055771.4
XM_005271453.1 Intron XP_005271510.1
XM_005271456.1 Intron XP_005271513.1
XM_005271457.1 Intron XP_005271514.1
XM_006718788.1 Intron XP_006718851.1
XM_006718794.1 Intron XP_006718857.1
XM_011542674.1 Intron XP_011540976.1
XM_011542690.1 Intron XP_011540992.1
XM_017017364.1 Intron XP_016872853.1
XM_017017365.1 Intron XP_016872854.1
XM_017017366.1 Intron XP_016872855.1
XM_017017367.1 Intron XP_016872856.1
XM_017017368.1 Intron XP_016872857.1
XM_017017369.1 Intron XP_016872858.1
XM_017017370.1 Intron XP_016872859.1
XM_017017371.1 Intron XP_016872860.1
XM_017017372.1 Intron XP_016872861.1
XM_017017373.1 Intron XP_016872862.1
XM_017017374.1 Intron XP_016872863.1
XM_017017375.1 Intron XP_016872864.1
XM_017017376.1 Intron XP_016872865.1
XM_017017377.1 Intron XP_016872866.1
XM_017017378.1 Intron XP_016872867.1
XM_017017379.1 Intron XP_016872868.1
XM_017017380.1 Intron XP_016872869.1
XM_017017381.1 Intron XP_016872870.1
XM_017017382.1 Intron XP_016872871.1
XM_017017383.1 Intron XP_016872872.1
XM_017017384.1 Intron XP_016872873.1
XM_017017385.1 Intron XP_016872874.1
XM_017017386.1 Intron XP_016872875.1

View Full Product Details