Product Details

SNP ID
rs112607377
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124919286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTGAGCATCCAGTATGTGCTA[C/G]GCACTCTGCTGGATGCTAGTAATAC
Phenotype
MIM: 611642 MIM: 611641
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HEPACAM PubMed Links
Additional Information
For this assay, SNP(s) [rs560243987] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HEPACAM
Gene Name
hepatic and glial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152722.4 37 UTR 3 NP_689935.2
XM_005271449.2 37 UTR 3 XP_005271506.1
XM_017017361.1 37 UTR 3 XP_016872850.1
Gene
HEPN1
Gene Name
hepatocellular carcinoma, down-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037558.2 37 UTR 5 NP_001032647.2

View Full Product Details