Product Details

SNP ID
rs117955196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130231840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAAGACCACAACGTAACATTA[A/C]AATTCCTGACCTTTCCAGTTGAGAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB44 PubMed Links

Gene Details

Gene
ZBTB44
Gene Name
zinc finger and BTB domain containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301098.1 4197 Intron NP_001288027.1
NM_001301099.1 4197 Intron NP_001288028.1
NM_014155.4 4197 UTR 3 NP_054874.3
XM_005271521.4 4197 UTR 3 XP_005271578.1
XM_005271522.4 4197 UTR 3 XP_005271579.1
XM_006718825.3 4197 UTR 3 XP_006718888.1
XM_006718826.3 4197 UTR 3 XP_006718889.1
XM_011542789.2 4197 Intron XP_011541091.1
XM_017017623.1 4197 UTR 3 XP_016873112.1
XM_017017624.1 4197 UTR 3 XP_016873113.1

View Full Product Details