Product Details

SNP ID
rs113292086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134153023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTCTTTATTCCTGGCGGGAGAC[C/T]GCACATTCCACTGCTGAGGCTGTGG
Phenotype
MIM: 606871 MIM: 609276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JAM3 PubMed Links

Gene Details

Gene
JAM3
Gene Name
junctional adhesion molecule 3
There are no transcripts associated with this gene.

Gene
NCAPD3
Gene Name
non-SMC condensin II complex subunit D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015261.2 5024 Missense Mutation NP_056076.1
XM_011542719.2 5024 Missense Mutation XP_011541021.1

View Full Product Details