Product Details

SNP ID
rs112393339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134154638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCGCCCCTCCTGGGTGGTGCT[A/G]TACCCAGTGCATAAGCTGCCACCAG
Phenotype
MIM: 606871 MIM: 609276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JAM3 PubMed Links

Gene Details

Gene
JAM3
Gene Name
junctional adhesion molecule 3
There are no transcripts associated with this gene.

Gene
NCAPD3
Gene Name
non-SMC condensin II complex subunit D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015261.2 Intron NP_056076.1
XM_011542719.2 Intron XP_011541021.1

View Full Product Details