Product Details

SNP ID
rs112423474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105041636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTTTGTTTCCATATCCTTTGAG[C/T]GTCTTCTAGAAAGCAAAGCTTGATT
Phenotype
MIM: 615680 MIM: 147678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD16 PubMed Links

Gene Details

Gene
CARD16
Gene Name
caspase recruitment domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017534.1 448 Missense Mutation ACT,GCT T120A NP_001017534.1
NM_052889.2 448 UTR 3 NP_443121.1
XM_011542583.2 448 Missense Mutation ACT,GCT T188A XP_011540885.1
Gene
CASP1
Gene Name
caspase 1
There are no transcripts associated with this gene.

View Full Product Details