Product Details

SNP ID
rs115924080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55440636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCAGCTTCCCCATTTGAGCGG[C/T]TGTCTCCTGCCCTCTTGCCCCAGGT
Phenotype
MIM: 612919 MIM: 611915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LANCL2 PubMed Links

Gene Details

Gene
LANCL2
Gene Name
LanC like 2
There are no transcripts associated with this gene.

Gene
VOPP1
Gene Name
vesicular, overexpressed in cancer, prosurvival protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284282.1 Intron NP_001271211.1
NM_001284283.1 Intron NP_001271212.1
NM_001284284.1 Intron NP_001271213.1
NM_001321242.1 Intron NP_001308171.1
NM_001321243.1 Intron NP_001308172.1
NM_001321244.1 Intron NP_001308173.1
NM_001321246.1 Intron NP_001308175.1
NM_001321247.1 Intron NP_001308176.1
NM_001321248.1 Intron NP_001308177.1
NM_001321249.1 Intron NP_001308178.1
NM_001321250.1 Intron NP_001308179.1
NM_001321251.1 Intron NP_001308180.1
NM_030796.4 Intron NP_110423.3
XM_011515539.1 Intron XP_011513841.1
XM_011515540.2 Intron XP_011513842.1
XM_011515541.2 Intron XP_011513843.1
XM_011515546.2 Intron XP_011513848.1

View Full Product Details