Product Details

SNP ID
rs111336486
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56060354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAGGACCAAATAAGCACACAC[C/T]CACTCAGATCAAAGATGCAGTGAGG
Phenotype
MIM: 104613 MIM: 172480 MIM: 607940
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCT6A PubMed Links
Additional Information
For this assay, SNP(s) [rs71015173] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CCT6A
Gene Name
chaperonin containing TCP1 subunit 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009186.1 Intron NP_001009186.1
NM_001762.3 Intron NP_001753.1
Gene
PSPH
Gene Name
phosphoserine phosphatase
There are no transcripts associated with this gene.

Gene
SNORA15
Gene Name
small nucleolar RNA, H/ACA box 15
There are no transcripts associated with this gene.

Gene
SUMF2
Gene Name
sulfatase modifying factor 2
There are no transcripts associated with this gene.

View Full Product Details