Product Details

SNP ID
rs116255634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:103086366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGTGGGGGTTAAGACTTCAAC[A/T]CAAATTTTGGGAGGGGCACATATCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GIN1 PubMed Links

Gene Details

Gene
GIN1
Gene Name
gypsy retrotransposon integrase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317954.1 2989 UTR 3 NP_001304883.1
NM_017676.2 2989 UTR 3 NP_060146.2
XM_005272028.3 2989 UTR 3 XP_005272085.1
XM_017009599.1 2989 Intron XP_016865088.1

View Full Product Details