Product Details

SNP ID
rs114368263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95556630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGATCACAACATCCCAGAAAC[C/T]TCCAAAGATGACTGAGATATATAGG
Phenotype
MIM: 610011 MIM: 614589
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSK PubMed Links

Gene Details

Gene
ARSK
Gene Name
arylsulfatase family member K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198150.2 Intron NP_937793.1
XM_005271904.3 Intron XP_005271961.1
Gene
TTC37
Gene Name
tetratricopeptide repeat domain 37
There are no transcripts associated with this gene.

View Full Product Details