Product Details

SNP ID
rs112713107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126870672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGAGTTCCTCTTGACCGAATG[G/T]AGGCCCAGCAAGGACGGCTGGTTAG
Phenotype
MIM: 613333
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MARCH3 PubMed Links

Gene Details

Gene
MARCH3
Gene Name
membrane associated ring-CH-type finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178450.4 1237 Silent Mutation CTA,CTC L241L NP_848545.1
XM_011543128.2 1237 Silent Mutation CTA,CTC L293L XP_011541430.1
XM_011543129.1 1237 Silent Mutation CTA,CTC L268L XP_011541431.1
XM_011543130.1 1237 Silent Mutation CTA,CTC L268L XP_011541432.1
XM_011543131.2 1237 Silent Mutation CTA,CTC L268L XP_011541433.1
XM_011543132.1 1237 Intron XP_011541434.1
XM_017009014.1 1237 Silent Mutation CTA,CTC L187L XP_016864503.1
XM_017009015.1 1237 Intron XP_016864504.1

View Full Product Details