Product Details

SNP ID
rs116135742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131178054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATTGATGTTATTCCATCTCTG[C/T]GATAACATGTTTCTGAGGAAGTTTT
Phenotype
MIM: 615831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYRM7 PubMed Links

Gene Details

Gene
LYRM7
Gene Name
LYR motif containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293735.1 Intron NP_001280664.1
NM_181705.3 Intron NP_859056.2

View Full Product Details