Product Details

SNP ID
rs113433326
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138946988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAGACACACAGCAGAAAGAGG[A/T]TGGCCTTCAGGTTTCCATTTAATGG
Phenotype
MIM: 608005
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
SIL1 PubMed Links

Gene Details

Gene
SIL1
Gene Name
SIL1 nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037633.1 1758 UTR 3 NP_001032722.1
NM_022464.4 1758 UTR 3 NP_071909.1
XM_011543570.2 1758 UTR 3 XP_011541872.1

View Full Product Details