Product Details

SNP ID
rs113408838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139342610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCATGCCCTGGAAAAAAGAAC[C/T]TTTGGGGTACTGGGAACCCTTACCC
Phenotype
MIM: 605604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAIP2 PubMed Links

Gene Details

Gene
PAIP2
Gene Name
poly(A) binding protein interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033112.2 761 UTR 5 NP_001028284.1
NM_016480.4 761 Intron NP_057564.3
XM_011543428.1 761 UTR 5 XP_011541730.1
XM_017009522.1 761 UTR 5 XP_016865011.1
XM_017009523.1 761 Intron XP_016865012.1

View Full Product Details