Product Details

SNP ID
rs115404031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:100536008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCACCCCCGTGGGCCTGAGATA[C/T]GCAGGCCGGTGATCTCCAGCAAGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM174A PubMed Links

Gene Details

Gene
FAM174A
Gene Name
family with sequence similarity 174 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198507.2 Intron NP_940909.1
XM_006714600.1 Intron XP_006714663.1

View Full Product Details