Product Details
- SNP ID
-
rs114397779
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:132595025 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCGTTTTGCTTTTTGACCTTGGC[A/G]ATGTCGGCCTCCATGCCGCTTGAGT
- Phenotype
-
MIM: 605211
MIM: 616533
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BARHL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs306547] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BARHL1
- Gene Name
- BarH like homeobox 1
There are no transcripts associated with this gene.
- Gene
- DDX31
- Gene Name
- DEAD-box helicase 31
View Full Product Details