Product Details

SNP ID
rs111352323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133356599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGCGGGCGCGTCGGCCATGAGC[A/G]AGTTGCCGGGCGACGTGCGGGCGTT
Phenotype
MIM: 185641 MIM: 185640 MIM: 185620 MIM: 185630 MIM: 185660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED22 PubMed Links

Gene Details

Gene
MED22
Gene Name
mediator complex subunit 22
There are no transcripts associated with this gene.

Gene
RPL7A
Gene Name
ribosomal protein L7a
There are no transcripts associated with this gene.

Gene
SNORD24
Gene Name
small nucleolar RNA, C/D box 24
There are no transcripts associated with this gene.

Gene
SNORD36A
Gene Name
small nucleolar RNA, C/D box 36A
There are no transcripts associated with this gene.

Gene
SNORD36B
Gene Name
small nucleolar RNA, C/D box 36B
There are no transcripts associated with this gene.

Gene
SNORD36C
Gene Name
small nucleolar RNA, C/D box 36C
There are no transcripts associated with this gene.

Gene
SURF1
Gene Name
surfeit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280787.1 55 Intron NP_001267716.1
NM_003172.3 55 Intron NP_003163.1
XM_011518942.1 55 Intron XP_011517244.1
Gene
SURF2
Gene Name
surfeit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278928.1 55 Missense Mutation AAG,GAG K3E NP_001265857.1
NM_017503.4 55 Missense Mutation AAG,GAG K3E NP_059973.4
Gene
SURF4
Gene Name
surfeit 4
There are no transcripts associated with this gene.

View Full Product Details