Product Details

SNP ID
rs116483979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8391854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGTCCGTGTGGCGAAAGCCTTC[C/T]GGGGGGAGGGAGAGGAAGTGCCTCT
Phenotype
MIM: 604198
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4999 PubMed Links

Gene Details

Gene
MIR4999
Gene Name
microRNA 4999
There are no transcripts associated with this gene.

Gene
RAB11B
Gene Name
RAB11B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004218.3 Intron NP_004209.2
Gene
RAB11B-AS1
Gene Name
RAB11B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details