Product Details

SNP ID
rs115395861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8065533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTACTCCGAGGAATAAGCCCT[A/G]TGCCCACCCCAGTTGCCCATTGCCA
Phenotype
MIM: 602565 MIM: 608529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL25 PubMed Links

Gene Details

Gene
CCL25
Gene Name
C-C motif chemokine ligand 25
There are no transcripts associated with this gene.

Gene
FBN3
Gene Name
fibrillin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321431.1 8956 UTR 3 NP_001308360.1
NM_032447.4 8956 UTR 3 NP_115823.3
XM_017027372.1 8956 UTR 3 XP_016882861.1
XM_017027373.1 8956 UTR 3 XP_016882862.1
XM_017027374.1 8956 UTR 3 XP_016882863.1
XM_017027375.1 8956 UTR 3 XP_016882864.1
XM_017027376.1 8956 UTR 3 XP_016882865.1
XM_017027377.1 8956 UTR 3 XP_016882866.1
XM_017027378.1 8956 UTR 3 XP_016882867.1
XM_017027379.1 8956 UTR 3 XP_016882868.1

View Full Product Details