Product Details

SNP ID
rs112031241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47626160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAGGTACTCCACTCTGACCAT[A/G]ATCCTTTCACTGCAATGAGGGGTAG
Phenotype
MIM: 300383 MIM: 311040 MIM: 313440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFP PubMed Links

Gene Details

Gene
CFP
Gene Name
complement factor properdin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145252.1 1268 Missense Mutation TCA,TTA S381L NP_001138724.1
NM_002621.2 1268 Missense Mutation TCA,TTA S381L NP_002612.1
XM_017029575.1 1268 Missense Mutation TCA,TTA S246L XP_016885064.1
Gene
ELK1
Gene Name
ELK1, ETS transcription factor
There are no transcripts associated with this gene.

Gene
SYN1
Gene Name
synapsin I
There are no transcripts associated with this gene.

View Full Product Details