Product Details

SNP ID
rs116316712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47846122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCAGAGTTCTTGGAGTCTCTG[C/T]GGAGTCCCTGGGCCAGATCTCACAG
Phenotype
MIM: 314998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF81 PubMed Links

Gene Details

Gene
ZNF81
Gene Name
zinc finger protein 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007137.3 104 UTR 5 NP_009068.2
XM_005272600.3 104 UTR 5 XP_005272657.1
XM_011543899.2 104 UTR 5 XP_011542201.1
XM_011543900.2 104 UTR 5 XP_011542202.1
XM_017029486.1 104 UTR 5 XP_016884975.1
XM_017029487.1 104 UTR 5 XP_016884976.1

View Full Product Details