Product Details

SNP ID
rs111262473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48195353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCGTACAAAGGCATCGTCTCC[A/G]TTCATGGCACCGGGAGCACTCTGTC
Phenotype
MIM: 300327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSX5 PubMed Links

Gene Details

Gene
SSX5
Gene Name
SSX family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021015.3 94 Intron NP_066295.3
NM_175723.1 94 Intron NP_783729.1
XM_011543949.2 94 Silent Mutation AAC,AAT N2N XP_011542251.1
XM_017029758.1 94 Silent Mutation AAC,AAT N2N XP_016885247.1

View Full Product Details