Product Details
- SNP ID
-
rs113588421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:40052340 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACGTTTGGAAAATTGCAGCGAAA[A/T]ATGCGGGAGGACATTTTCAATTTCT
- Phenotype
-
MIM: 300485
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BCOR
PubMed Links
Gene Details
- Gene
- BCOR
- Gene Name
- BCL6 corepressor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001123383.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1645I |
NP_001116855.1 |
NM_001123384.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1627I |
NP_001116856.1 |
NM_001123385.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
NP_001116857.1 |
NM_017745.5 |
5461 |
Silent Mutation |
ATA,ATT |
I1645I |
NP_060215.4 |
XM_005272616.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_005272673.1 |
XM_005272618.3 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_005272675.1 |
XM_005272619.3 |
5461 |
Silent Mutation |
ATA,ATT |
I1661I |
XP_005272676.1 |
XM_005272620.3 |
5461 |
Silent Mutation |
ATA,ATT |
I1627I |
XP_005272677.1 |
XM_006724536.3 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_006724599.1 |
XM_011543929.2 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_011542231.1 |
XM_011543930.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_011542232.1 |
XM_011543931.2 |
5461 |
Silent Mutation |
ATA,ATT |
I1679I |
XP_011542233.1 |
XM_017029615.1 |
5461 |
Silent Mutation |
ATA,ATT |
I1645I |
XP_016885104.1 |
XM_017029616.1 |
5461 |
UTR 3 |
|
|
XP_016885105.1 |
View Full Product Details