Product Details

SNP ID
rs111493766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40654501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCCATGTCATAAATGATTGGA[A/G]CTATAATACTAACAGGTTCTTGGGG
Phenotype
MIM: 300182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf38 PubMed Links

Gene Details

Gene
CXorf38
Gene Name
chromosome X open reading frame 38
There are no transcripts associated with this gene.

Gene
MED14
Gene Name
mediator complex subunit 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004229.3 4204 Missense Mutation GCT,GTT A1385V NP_004220.2
XM_005272701.2 4204 Missense Mutation GCT,GTT A1269V XP_005272758.1
XM_011544000.2 4204 Missense Mutation GCT,GTT A1269V XP_011542302.1
XM_017029962.1 4204 Missense Mutation GCT,GTT A1269V XP_016885451.1

View Full Product Details