Product Details

SNP ID
rs112733631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43666428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACCACATCCATTCCAGCAGCAA[A/G]TCCTATTGGCTGTAGCCTCAAAATA
Phenotype
MIM: 309850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAOA PubMed Links

Gene Details

Gene
MAOA
Gene Name
monoamine oxidase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000240.3 Intron NP_000231.1
NM_001270458.1 Intron NP_001257387.1

View Full Product Details