Product Details

SNP ID
rs111398282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133325983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTTGGCTGCGCGGTAGCCGTC[A/C]CCCCAGGCCGCCGGCGTCTCCGTGC
Phenotype
MIM: 604647
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALY PubMed Links

Gene Details

Gene
CALY
Gene Name
calcyon neuron specific vesicular protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321617.1 1800 Silent Mutation GGG,GGT G84G NP_001308546.1
NM_015722.3 1800 Silent Mutation GGG,GGT G166G NP_056537.1
XM_005252685.4 1800 Silent Mutation GGG,GGT G199G XP_005252742.2
XM_011539836.2 1800 Silent Mutation GGG,GGT G199G XP_011538138.1
XM_011539838.2 1800 Silent Mutation GGG,GGT G79G XP_011538140.1

View Full Product Details