Product Details

SNP ID
rs111549370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119831614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACAGACACCTGGTTTGAAACA[A/C]AGAAACAAATTTAAGTCTAGAGCTG
Phenotype
MIM: 609389 MIM: 610909
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INPP5F PubMed Links

Gene Details

Gene
INPP5F
Gene Name
inositol polyphosphate-5-phosphatase F
There are no transcripts associated with this gene.

Gene
MCMBP
Gene Name
minichromosome maintenance complex binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256378.1 Intron NP_001243307.1
NM_001256379.1 Intron NP_001243308.1
NM_024834.3 Intron NP_079110.1
XM_005270157.4 Intron XP_005270214.1
XM_011540170.1 Intron XP_011538472.1
XM_017016663.1 Intron XP_016872152.1
XM_017016664.1 Intron XP_016872153.1

View Full Product Details