Product Details

SNP ID
rs111718112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89402482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACACCTGAAAGGCCAGAATGA[A/G]GAAGCCCTGAAGAGCTTAAAAGAAG
Phenotype
MIM: 147690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFIT1 PubMed Links

Gene Details

Gene
IFIT1
Gene Name
interferon induced protein with tetratricopeptide repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270927.1 483 Silent Mutation GAA,GAG E69E NP_001257856.1
NM_001270928.1 483 Silent Mutation GAA,GAG E38E NP_001257857.1
NM_001270929.1 483 Silent Mutation GAA,GAG E38E NP_001257858.1
NM_001270930.1 483 Silent Mutation GAA,GAG E38E NP_001257859.1
NM_001548.4 483 Silent Mutation GAA,GAG E69E NP_001539.3

View Full Product Details