Product Details

SNP ID
rs112344790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23439494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCGTCTGCACCCACTACCCC[A/G]CCGGGGCCCCGGGTCCCACGGCCGC
Phenotype
MIM: 612022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD1 PubMed Links

Gene Details

Gene
OTUD1
Gene Name
OTU deubiquitinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145373.2 226 Missense Mutation ACC,GCC T13A NP_001138845.1

View Full Product Details