Product Details
- SNP ID
-
rs112423286
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:32907119 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGATTCTTGAAATTATTAATAG[C/G]ACTCTTGTAAATCGGATTTTCTTGC
- Phenotype
-
MIM: 135630
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ITGB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80132666] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITGB1
- Gene Name
- integrin subunit beta 1
- Gene
- SNORA86
- Gene Name
- small nucleolar RNA, H/ACA box 86
There are no transcripts associated with this gene.
View Full Product Details