Product Details

SNP ID
rs112461952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110502328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGAAGAAGCCACTTCAGGAGC[A/C]TGTCTTCTCGCCAATTAATGAAATT
Phenotype
MIM: 603069
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DUSP5 PubMed Links

Gene Details

Gene
DUSP5
Gene Name
dual specificity phosphatase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004419.3 Intron NP_004410.3

View Full Product Details