Product Details
- SNP ID
-
rs113570783
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:101010532 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTACCTGGAGACTTGCCTTGACCC[C/T]GGCTGCTGCGGCTGCGGCTGCGGCT
- Phenotype
-
MIM: 610454
MIM: 612971
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LZTS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200896335] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LZTS2
- Gene Name
- leucine zipper tumor suppressor 2
There are no transcripts associated with this gene.
- Gene
- PDZD7
- Gene Name
- PDZ domain containing 7
View Full Product Details