Product Details

SNP ID
rs113848933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100488080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCCAGACGCTGGGCTGCCTCTT[A/C]CAGCTTCCTTTTTGTCTTCTGCAGG
Phenotype
MIM: 610258 MIM: 601396
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC31B PubMed Links

Gene Details

Gene
SEC31B
Gene Name
SEC31 homolog B, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015490.3 3409 Nonsense Mutation GAA,TAA E1103* NP_056305.1
Gene
WNT8B
Gene Name
Wnt family member 8B
There are no transcripts associated with this gene.

View Full Product Details