Product Details

SNP ID
rs114656691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86762838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGCTTTCCTGAGAAAGGTAGCC[C/T]GCAGAGTGAGACCTTAGGCAAGAAA
Phenotype
MIM: 601299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPR1A PubMed Links

Gene Details

Gene
BMPR1A
Gene Name
bone morphogenetic protein receptor type 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004329.2 Intron NP_004320.2
XM_011540103.2 Intron XP_011538405.1
XM_011540104.2 Intron XP_011538406.1

View Full Product Details