Product Details
- SNP ID
-
rs114686645
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:71216897 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATAGACAGTGCTGGTACCTCCCT[A/C]CCCCAGGGCCTGCAGGCAGCCCGTC
- Phenotype
-
MIM: 607870
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UNC5B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140272889] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UNC5B
- Gene Name
- unc-5 netrin receptor B
- Gene
- UNC5B-AS1
- Gene Name
- UNC5B antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details