Product Details

SNP ID
rs114702481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45303720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGGAGACGATGAAGCCATAGG[A/G]CGCGATGGTCATCAGGAAGTTCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13A1 PubMed Links

Gene Details

Gene
OR13A1
Gene Name
olfactory receptor family 13 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004297.2 4785 Missense Mutation CCC,TCC P235S NP_001004297.2
XM_011540147.2 4785 Missense Mutation CCC,TCC P235S XP_011538449.1
XM_017016632.1 4785 Missense Mutation CCC,TCC P235S XP_016872121.1
XM_017016633.1 4785 Missense Mutation CCC,TCC P235S XP_016872122.1
XM_017016634.1 4785 Missense Mutation CCC,TCC P235S XP_016872123.1

View Full Product Details