Product Details

SNP ID
rs114806719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100235554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATCTGCCCTAAGACCATGGGT[C/T]CTCTGGGGCAAGGACTGTTTACAAT
Phenotype
MIM: 600664 MIM: 616120 MIM: 611330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHUK PubMed Links

Gene Details

Gene
CHUK
Gene Name
conserved helix-loop-helix ubiquitous kinase
There are no transcripts associated with this gene.

Gene
CWF19L1
Gene Name
CWF19-like 1, cell cycle control (S. pombe)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303404.1 Intron NP_001290333.1
NM_001303405.1 Intron NP_001290334.1
NM_001303406.1 Intron NP_001290335.1
NM_001303407.1 Intron NP_001290336.1
NM_018294.5 Intron NP_060764.3
Gene
SNORA12
Gene Name
small nucleolar RNA, H/ACA box 12
There are no transcripts associated with this gene.

View Full Product Details