Product Details

SNP ID
rs114934821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88180048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTTGTATACAGTATGTTCTTA[C/T]GGAAACCACACTTAGAATGCAGAGT
Phenotype
MIM: 609360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNLS PubMed Links

Gene Details

Gene
RNLS
Gene Name
renalase, FAD dependent amine oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031709.2 4554 Intron NP_001026879.2
NM_018363.3 4554 Intron NP_060833.1
XM_005269946.1 4554 Intron XP_005270003.1
XM_005269947.1 4554 Intron XP_005270004.1
XM_005269948.2 4554 Intron XP_005270005.1
XM_005269949.4 4554 Intron XP_005270006.1
XM_005269950.3 4554 Intron XP_005270007.1
XM_011539924.2 4554 UTR 3 XP_011538226.1
XM_011539927.2 4554 Intron XP_011538229.1
XM_017016380.1 4554 Intron XP_016871869.1
XM_017016381.1 4554 Intron XP_016871870.1
XM_017016382.1 4554 UTR 3 XP_016871871.1
XM_017016383.1 4554 Intron XP_016871872.1
XM_017016384.1 4554 Intron XP_016871873.1
XM_017016385.1 4554 Intron XP_016871874.1

View Full Product Details